Learn More
SLC3A2 Mouse anti-Human, Clone: 44D7, Abnova™
Mouse monoclonal antibody raised against human SLC3A2.
Manufacturer:  Abnova Corporation MAB12567
Catalog No. 89-934-938
Add to cart
Description
Description
This gene is a member of the solute carrier family and encodes a cell surface, transmembrane protein with an alpha amylase domain. The protein exists as the heavy chain of a heterodimer, covalently bound through di-sulfide bonds to one of several possible light chains. It associates with integrins and mediates integrin-dependent signaling related to normal cell growth and tumorigenesis. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. [provided by RefSeq]
Specifications
Specifications
SLC3A2
44D7
Mouse monoclonal antibody raised against human SLC3A2.
In PBS (0.09% sodium azide).
4F2/4F2HC/4T2HC/CD98/CD98HC/MDU1/NACAE
Mouse
IgG1
50 ug
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (PFA fixed), Immunoprecipitation
Unconjugated
Flow Cytometry (1:50-1:200) Immunohistochemistry (Frozen sections) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (15 ug/mL) Immunoprecipitation The optimal working dilution should be determined by the end user.
Liquid
SLC3A2
HOON pre-B leukaemia cell line
Protein G purification
RUO
Primary
6520