Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC43A2 Rabbit anti-Human, Alexa Fluor 647, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP306013AF647
Description
SLC43A2 Polyclonal specifically detects SLC43A2 in Human samples. It is validated for Western Blot.Specifications
SLC43A2 | |
Polyclonal | |
Western Blot | |
FLJ23848, large neutral amino acids transporter small subunit 4, LAT4amino acid transporter, L-type amino acid transporter 4, MGC34680, Solute carrier family 43 member 2, solute carrier family 43, member 2 | |
Produced in rabbits immunized with E. coli-derived Human SLC43A2 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
Alexa Fluor 647 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
124935 | |
Store at 4°C in the dark. | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title