Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLC45A2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA599036
Description
SLC45A2 Polyclonal Antibody for Western Blot, ELISA
SLC45A2 is a melanocyte differentiation antigen that is expressed in a high percentage of melanoma cell lines. A similar sequence gene in medaka, 'B, ' encodes a transporter that mediates melanin synthesis. Mutations in this gene are a cause of oculocutaneous albinism type 4. The protein encoded by this gene encodes a melanocyte differentiation antigen that is expressed in a high percentage of melanoma cell lines. A similar sequence gene in medaka, 'B, ' encodes a transporter that mediates melanin synthesis. Mutations in this gene are a cause of oculocutaneous albinism type 4. Alternative splicing results in multiple transcript variants encoding different isoforms.
Specifications
SLC45A2 | |
Polyclonal | |
Unconjugated | |
SLC45A2 | |
1A1; AIM1; Aim-1; B/AIM-1-like protein; blanc-sale; bls; Dbr; dominant brown; Matp; Melanoma antigen AIM1; membrane associated transport protein; membrane associated transporter protein; membrane-associated transporter protein; Oca4; protein AIM-1; Protein underwhite; SHEP5; Slc45a2; Solute carrier family 45 member 2; solute carrier family 45, member 2; underwhite; uw | |
Rabbit | |
Protein G | |
RUO | |
51151 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Western Blot | |
3.31 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q9UMX9 | |
SLC45A2 | |
Recombinant Human Membrane-associated transporter protein (1-46AA). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction