Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC45A2 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$152.22 - $436.00
Specifications
Antigen | SLC45A2 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP15978620
|
Novus Biologicals
NBP15978620UL |
20 μL |
Each for $152.22
|
|
NBP159786
|
Novus Biologicals
NBP159786 |
100 μL |
Each for $436.00
|
|
Description
SLC45A2 Polyclonal specifically detects SLC45A2 in Human samples. It is validated for Western Blot.Specifications
SLC45A2 | |
Polyclonal | |
Rabbit | |
Breast Cancer | |
Q6P2P0 | |
51151 | |
IgG | |
Affinity Purified |
Western Blot | |
Unconjugated | |
RUO | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
AIM-1, AIM11A1, MATPmembrane associated transporter, Melanoma antigen AIM1, membrane-associated transporter protein, Protein AIM-1, SHEP5, Solute carrier family 45 member 2, solute carrier family 45, member 2, underwhite | |
SLC45A2 | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title