Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC4A7 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA550525

Catalog No. PA550525


Add to cart

Description

Description

The antibody detects endogenous levels of total SLC4A7 protein.

SLC4A7 regulates intracellular pH and may also play a role in bicarbonate salvage in secretory epithelia. Additionally, SLC4A7 may also have associated sodium channel activity.
Specifications

Specifications

SLC4A7
Polyclonal
Unconjugated
SLC4A7
Liquid
SLC4A7
Synthetic peptide corresponding to a region derived from internal residues of human Solute carrier family 4, sodium bicarbonate cotransporter, member 7
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin)
1.8 mg/mL
PBS with 50% glycerol and 0.05% sodium azide; pH 7.3
Q9Y6M7
BT, NBC2, NBC2B, NBC3, NBCn1, SBC2, SLC4A6
Rabbit
IgG
100 μL
-20°C
Primary
9497
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit