Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC5A11 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | SLC5A11 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP15963520
![]() |
Novus Biologicals
NBP15963520UL |
20 μL |
Each for $206.00
|
|
|||||
NBP159635
![]() |
Novus Biologicals
NBP159635 |
100 μL |
Each for $487.50
|
|
|||||
Description
SLC5A11 Polyclonal specifically detects SLC5A11 in Human samples. It is validated for Western Blot.Specifications
SLC5A11 | |
Polyclonal | |
Rabbit | |
Q8WWX8 | |
115584 | |
IgG | |
74 kDa |
Western Blot | |
Unconjugated | |
RUO | |
KST1RKST1, Na(+)/myo-inositol cotransporter 2, na+/myo-inositol cotransporter 2, putative sodium-coupled cotransporter RKST1, SGLT6, SLGTX, SMIT2homolog of rabbit KST1, Sodium/glucose cotransporter KST1, sodium/myo-inositol cotransporter 2, Sodium-dependent glucose cotransporter, solute carrier family 5 (sodium/glucose cotransporter), member 11, Solute carrier family 5 member 11 | |
SLC5A11 | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title