Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC5A11 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA550764

Catalog No. PA550764


Only null left
Add to Cart

Description

Description

The antibody detects endogenous levels of total SMIT2 / SLC5A11 protein.

Cotransporters, such as SLC5A11, represent a major class of proteins that make use of ion gradients to drive active transport for the cellular accumulation of nutrients, neurotransmitters, osmolytes, and ions Roll et al. (2002).
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SLC5A11
Polyclonal
Unconjugated
SLC5A11
2010013B02Rik; homolog of rabbit KST1; hypothetical protein LOC539084; KST1; Na(+)/myo-inositol cotransporter 2; na+/myo-inositol cotransporter 2; Na+/myo-inositol cotransporter SMIT2; putative sodium-coupled cotransporter RKST1; R SGLT6; RGD:621410}; RKST1; RKST2; SGLT6; Slc5a10; slc5a11; slc5a11 {ECO:0000312; SLGTX; SMIT2; sodium/glucose cotransporter KST1; Sodium/myo-inositol cotransporter 2; sodium/myo-inositol transporter 2; sodium-cotransporter rkST1; sodium-coupled cotransporter KST1; sodium-dependent glucose cotransporter; sodium-glucose cotransporter-like protein; solute carrier family 5 (sodium/glucose cotransporter), member 10; solute carrier family 5 (sodium/glucose cotransporter), member 11; solute carrier family 5 (sodium/inositol cotransporter), member 11; solute carrier family 5 member 11; zgc:92080
Rabbit
Antigen Affinity Chromatography
RUO
115584, 233836, 252854
-20°C
Liquid
Immunohistochemistry (Paraffin)
2.2 mg/mL
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Q8K0E3, Q8WWX8, Q9Z1F2
SLC5A11
Synthetic peptide corresponding to a region derived from internal residues of human solute carrier family 5 (sodium/inositol cotransporter), member 11.
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.