Learn More
SLC5A3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 216281AP
Catalog No. 50-173-3521
Add to cart
Description
Description
Prevents intracellular accumulation of high concentrations of myo-inositol (an osmolyte) that result in impairment of cellular function.
Specifications
Specifications
SLC5A3
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P53794, Q9JKZ2
SLC5A3, SMIT, SMIT2
Rabbit
IgG
150 μL
-20°C
Primary
114507, 53881, 6526
Western Blot
Unconjugated
SLC5A3
Liquid
Slc5a3
SLC5A3 Fusion Protein Ag16218
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat