Learn More
SLC5A5/Sodium Iodide Symporter Mouse anti-Human, Alexa Fluor 750, Clone: 902320, R&D Systems™
Mouse MonoclonalAntibody
Manufacturer:  R&D Systems FAB8367S100UG
Catalog No. FAB8367S100
Add to cart
This item is not returnable. View return policy
Description
Description
SLC5A5/Sodium Iodide Symporter Monoclonal antibody specifically detects SLC5A5/Sodium Iodide Symporter in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
SLC5A5/Sodium Iodide Symporter
Monoclonal
0.2 mg/mL
Flow Cytometry 0.25-1 ug/10^6 cells
Q92911
SLC5A5
NS0 mouse myeloma cell line transfected with human SLC5A5 Accession # Q92911
100 ug
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Monoclonal
Stains human SLC5A5 transfectants but not irrelevant transfectants in flow cytometry.
Flow Cytometry
902320
Alexa Fluor 750
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Na(+)/I(-) cotransporter, Na(+)/I(-) symporter, NIS, NISNa(+)/I(-)-symporter, sodium/iodide cotransporter, Sodium-iodide symporter, solute carrier family 5 (sodium iodide symporter), member 5, Solute carrier family 5 member 5, TDH1
Mouse
IgG2a
RUO
Primary
6528
Human
SDS
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)