Learn More
SLC5A5/Sodium Iodide Symporter Mouse anti-Human, Mouse, Rat, DyLight 350, Clone: 14F, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259341UV
Catalog No. NBP259341UV
Add to cart
Description
Description
SLC5A5/Sodium Iodide Symporter Monoclonal antibody specifically detects SLC5A5/Sodium Iodide Symporter in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
Specifications
SLC5A5/Sodium Iodide Symporter
14F
Western Blot, Immunohistochemistry
Purified
SLC5A5
Mannose binding protein hNIS fusion (AA468-643)
Protein G purified
Store at 4C in the dark.
Monoclonal
Detects 97kDa, non-glycosylated version at 68kDa. Other minor bands associated with hNIS at 160kDa, and degradation products at 30 kDa, and 15kDa.
Western Blot, Immunohistochemistry
DyLight 350
50mM Sodium Borate with 0.05% Sodium Azide
Na(+)/I(-) cotransporter, Na(+)/I(-) symporter, NISNa(+)/I(-)-symporter, sodium/iodide cotransporter, Sodium-iodide symporter, solute carrier family 5 (sodium iodide symporter), member 5, Solute carrier family 5 member 5, TDH1
Mouse
IgG1
100 ul
Primary
6528
Human, Mouse, Rat