Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLC5A8 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113782
Description
Immunogen sequence: CNSTYNETNL MTTTEMPFTT SVFQIYNVQR TPLMDNWYSL SYLYFSTVGT For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Human: 100%.
SLC5A8 has been shown to transport iodide by a passive mechanism (Rodriguez et al., 2002 ) and monocarboxylates and short-chain fatty acids by a sodium-coupled mechanism (Gopal et al., 2004 ).
Specifications
SLC5A8 | |
Polyclonal | |
Unconjugated | |
Slc5a8 | |
AIT; Apical iodide transporter; apical iodide transporter , putative; electrogenic sodium monocarboxylate cotransporter; RGD1564146; Slc5a8; SMCT; SMCT1; Sodium iodide-related cotransporter; sodium-coupled monocarboxylate transporter 1; solute carrier family 5 (iodide transporter), member 8; solute carrier family 5 (sodium/monocarboxylate cotransporter), member 8; solute carrier family 5 member 8 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
160728 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8N695 | |
Slc5a8 | |
Synthetic peptide directed towards the C-terminal region of Human SLC5A8 (aa 480-529). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction