Learn More
SLC5A8/SMCT1 Mouse anti-Human, Alexa Fluor 594, Clone: 903502, R&D Systems™
Mouse MonoclonalAntibody
Manufacturer:  R&D Systems FAB8398T100UG
Catalog No. FAB8398T100
Add to cart
This item is not returnable. View return policy
Description
Description
SLC5A8/SMCT1 Monoclonal antibody specifically detects SLC5A8/SMCT1 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
SLC5A8/SMCT1
Monoclonal
0.2 mg/mL
Flow Cytometry 0.25-1 ug/10^6 cells
NP_666018
SLC5A8
NS0 mouse myeloma cell line transfected with human SLC5A8/SMCT1 Met1-Leu610 Accession # NP_666018
100 ug
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Monoclonal
Detects HEK293 human embryonic kidney cell line transfected with human SLC5A8/SMCT1 by Flow Cytometry. Does not detect untransfected or irrelevant transfected HEK293 cells.
Flow Cytometry
903502
Alexa Fluor 594
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
AIT, AITSolute carrier family 5 member 8, Apical iodide transporter, Electrogenic sodium monocarboxylate cotransporter, MGC125354, SMCT, SMCTSMCT1, Sodium iodide-related cotransporter, sodium-coupled monocarboxylate transporter 1, solute carrier family 5 (iodide transporter), member 8
Mouse
IgG1
RUO
Primary
160728
Human
SDS
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)