Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLC6A13 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA568331
Description
Predicted to react with Mouse and Rat samples.
Slc6a13 catalyzes sodium and chloride dependent transport of gamma-aminobutyric acid (GABA) to terminate GABA mediated synaptic activity. Slc6a13 is also a high-affinity gamma-aminobutyric acid (GABA) transporter, terminates biological actions of GABA by reuptake into cell, and its reuptake function utilizes sodium and chloride concentration gradients. Further, Slc6a13 may also aid in beta-alanine transport. Diseases associated with SLC6A13 include Traumatic Glaucoma and Neovascular Glaucoma.
Specifications
SLC6A13 | |
Polyclonal | |
Unconjugated | |
Slc6a13 | |
GABA transport protein; GABA transporter 2; Gabt2; Gabt3; Gabt4; gamma-aminobutyric acid (GABA-A) transporter 3; GAT2; GAT-2; GAT3; Gat-3; Slc6a13; Sodium- and chloride-dependent GABA transporter 2; sodium- and chloride-dependent GABA transporter 3; solute carrier family 6 (neurotransmitter transporter), member 13; solute carrier family 6 (neurotransmitter transporter, GABA), member 13; solute carrier family 6 member 13 | |
Rabbit | |
Affinity chromatography | |
RUO | |
14412, 171163, 6540 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P31646, P31649, Q9NSD5 | |
Slc6a13 | |
Synthesized peptide derived from human SLC6A13. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction