Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLC6A14 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA542452
Description
Peptide sequence: VFAGFAIFSI LGHMAHISGK EVSQVVKSGF DLAFIAYPEA LAQLPGGPFW Sequence homology: Cow: 93%; Dog: 100%; Guinea Pig: 86%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%.
This gene encodes a member of the solute carrier family 6. Members of this family are sodium and chloride dependent neurotransmitter transporters. The encoded protein transports both neutral and cationic amino acids.
Specifications
SLC6A14 | |
Polyclonal | |
Unconjugated | |
SLC6A14 | |
1110007A17Rik; 9030613J17Rik; amino acid transporter ATB0+; amino acid transporter B0; amino acid transporter B0+; ATB(0+); Atb0+; ATB0plus; BMIQ11; carrier protein; CATB0plus; colonic system B0+ amino acid transporter; colonic system B0+ amino acid transporter CATB0+; LOW QUALITY PROTEIN: sodium- and chloride-dependent neutral and basic amino acid transporter B(0+); neutral cationic amino acid transporter B0,+; SLC6A14; Sodium- and chloride-dependent neutral and basic amino acid transporter B(0+); solute carrier family 6 (amino acid transporter), member 14; solute carrier family 6 (neurotransmitter transporter), member 14; solute carrier family 6 member 14 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
11254 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9UN76 | |
SLC6A14 | |
Synthetic peptide directed towards the middle region of human SLC6A14. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction