Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC6A15 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP159653
Description
SLC6A15 Polyclonal specifically detects SLC6A15 in Human samples. It is validated for Western Blot.Specifications
SLC6A15 | |
Polyclonal | |
Western Blot 1.0 ug/ml | |
Q9H2J7 | |
SLC6A15 | |
Synthetic peptides corresponding to SLC6A15 (solute carrier family 6 (neutral amino acid transporter), member 15) The peptide sequence was selected from the middle region of SLC6A15)(50ug). Peptide sequence YISPLMLLSLLIASVVNMGLSPPGYNAWIEDKASEEFLSYPTWGLVVCVS The peptide sequence for this immunogen was taken from within the described region. | |
Affinity Purified | |
RUO | |
55117 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
B0AT2, DKFZp761I0921, FLJ10316, homolog of rat orphan transporter v7-3, hv7-3, MGC87066, NTT73orphan sodium- and chloride-dependent neurotransmitter transporter NTT73, orphan transporter v7-3, SBAT1, Sodium- and chloride-dependent neurotransmitter transporter NTT73, sodium/chloride dependent neurotransmitter transporter Homo sapiens orphanneurotransmitter transporter NTT7, Sodium-coupled branched-chain amino-acid transporter 1, solute carrier family 6 (neurotransmitter transporter), member 15, solute carrier family 6 (neutral amino acid transporter), member 15, Solute carrier family 6 member 15, solute carrier family 6, member 15, Transporter v7-3, V7-3 | |
Rabbit | |
82 kDa | |
100 μL | |
Primary | |
Rat 85%. | |
Human, Mouse, Rat, Porcine, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title