Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC6A15 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
$482.50
Specifications
Antigen | SLC6A15 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP159653
|
Novus Biologicals
NBP159653 |
100 μL |
Each of 1 for $482.50
|
|
|||||
Description
SLC6A15 Polyclonal specifically detects SLC6A15 in Human samples. It is validated for Western Blot.Specifications
SLC6A15 | |
Polyclonal | |
Rabbit | |
Q9H2J7 | |
55117 | |
IgG | |
82 kDa |
Western Blot | |
Unconjugated | |
RUO | |
B0AT2, DKFZp761I0921, FLJ10316, homolog of rat orphan transporter v7-3, hv7-3, MGC87066, NTT73orphan sodium- and chloride-dependent neurotransmitter transporter NTT73, orphan transporter v7-3, SBAT1, Sodium- and chloride-dependent neurotransmitter transporter NTT73, sodium/chloride dependent neurotransmitter transporter Homo sapiens orphanneurotransmitter transporter NTT7, Sodium-coupled branched-chain amino-acid transporter 1, solute carrier family 6 (neurotransmitter transporter), member 15, solute carrier family 6 (neutral amino acid transporter), member 15, Solute carrier family 6 member 15, solute carrier family 6, member 15, Transporter v7-3, V7-3 | |
SLC6A15 | |
Primary |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title