Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC6A17 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548587

Catalog No. PA548587

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Predicted to react with mouse and rat based on sequence homology.

Functions as a sodium-dependent vesicular transporter selective for proline, glycine, leucine and alanine. In contrast to other members of this neurotransmitter transporter family, does not appear to be chloride-dependent.
Specifications

Specifications

SLC6A17
Polyclonal
PBS with 0.09% sodium azide
Q9H1V8
Sodium-dependent neutral amino acid transporter SLC6A17, Sodium-dependent neurotransmitter transporter NTT4, Solute carrier family 6 member 17, SLC6A17, NTT4
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
388662
Western Blot
Unconjugated
SLC6A17
Liquid
SLC6A17
KLH conjugated synthetic peptide between 647-675 amino acids from the C-terminal region of human SLC6A17.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit