Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLC6A20 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5104153
Description
Antibody detects endogenous levels of total SLC6A20.
Mediates the calcium-dependent uptake of imino acids such as L-proline, N-methyl-L-proline and pipecolate as well as N-methylated amino acids. Involved in the transport of glycine.
Specifications
SLC6A20 | |
Polyclonal | |
Unconjugated | |
SLC6A20 | |
A730081N20Rik; AU022428; IMINO-B; neurotransmitter transporter RB21A; orphan transporter XT3; rB21a; SIT1; Slc6a20; Slc6a20a; sodium- and chloride-dependent transporter XTRP3; Sodium- and chloride-dependent transporter XTRP3A; sodium/imino-acid transporter 1; solute carrier family 6 (neurotransmitter transporter), member 20; solute carrier family 6 (neurotransmitter transporter), member 20A; solute carrier family 6 (proline IMINO transporter), member 20; solute carrier family 6 member 20; Solute carrier family 6 member 20A; solute carrier family 6, member 20; transporter rB21A; Transporter rB21A homolog; X transporter protein 3; X transporter protein 3 similar 1; XT3; Xt3s1; Xtm3s1; Xtrp3; Xtrp3s1 | |
Rabbit | |
Affinity chromatography | |
RUO | |
102680, 113918, 54716 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q64093, Q8VDB9, Q9NP91 | |
SLC6A20, Slc6a20a | |
A synthesized peptide derived from human SLC6A20(Accession Q9NP91), corresponding to amino acid residues T246-S296. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction