Learn More
SLC6A3/DAT1 Mouse anti-Mouse, Rat, Human, Alexa Fluor™ 750, Clone: mAb16, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP222164AF750
Catalog No. NP222164A70
Add to cart
Description
Description
SLC6A3/DAT1 Monoclonal antibody specifically detects SLC6A3/DAT1 in Mouse,Rat,Human samples. It is validated for Western Blot,ELISA,Immunohistochemistry,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Paraffin).
Specifications
Specifications
SLC6A3/DAT1
mAb16
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
DA transporter, DATDAT1Solute carrier family 6 member 3, sodium-dependent dopamine transporter, solute carrier family 6 (neurotransmitter transporter, dopamine), member 3
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
Human, Mouse, Rat
ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
SLC6A3
Synthetic peptide corresponding to the N-terminus of rat DAT1. [UniProtNo. P23977]
0.1 mL
Primary
6531