Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC7A10 Rabbit anti-Mouse, Rat, FITC, Polyclonal, FabGennix
Rabbit Polyclonal Antibody
Supplier: Fabgennix Inc ASC1FITC
Description
SLC7A10 Polyclonal antibody specifically detects SLC7A10 in Mouse, Rat samples. It is validated for ELISA, Immunocytochemistry, Immunohistochemistry, Immunomicroscopy, Immunoprecipitation, Western BlotSpecifications
SLC7A10 | |
Polyclonal | |
FITC | |
SLC7A10 | |
AL024237; ASC1; Asc-1; Asc-type amino acid transporter 1; D- and L-serine transporter Asc-1; D7Bwg0847e; D-serine transporter; HASC-1; SLC7A10; solute carrier family 7 (cationic amino acid transporter, y+ system), member 10; solute carrier family 7 (neutral amino acid transporter light chain, asc system), member 10; solute carrier family 7 member 10; solute carrier family 7, (cationic amino acid transporter, y+ system) member 10; solute carrier family 7, (neutral amino acid transporter, y+ system) member 10 | |
Synthetic peptide taken within amino acid region 1-50 on rat Asc-type amino acid transporter 1 protein. | |
200 μL | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |
ELISA, Immunocytochemistry, Immunohistochemistry, Immunomicroscopy, Immunoprecipitation, Western Blot | |
0.5-1.5 mg/mL | |
proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8 | |
P63115, P63116 | |
Rabbit | |
Affinity chromatography | |
RUO | |
114518, 53896 | |
-20° C, store in dark | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction