Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC7A13 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP31100925UL
Description
SLC7A13 Polyclonal specifically detects SLC7A13 in Human samples. It is validated for Western Blot.Specifications
SLC7A13 | |
Polyclonal | |
Western Blot 1.0 ug/ml | |
solute carrier family 7 member 13, AGT1, AGT-1, solute carrier family 7 (anionic amino acid transporter), member 13, XAT2 | |
This SLC7A13 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 178-208 amino acids from the Central region of human SLC7A13. (NP_620172.2). | |
25 μg | |
Primary | |
Human | |
Purified |
Western Blot | |
Unconjugated | |
PBS buffer, 2% sucrose | |
Rabbit | |
Affinity purified | |
RUO | |
157724 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title