Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC7A8 Mouse anti-Human, Clone: OTI4H10, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA524976

Catalog No. PIMA524976


Add to cart

Description

Description

SLC7A8 is sodium-independent, high-affinity transport of large neutral amino acids. It has higher affinity for L-phenylalanine than LAT1 but lower affinity for glutamine and serine. L-alanine is transported at physiological concentrations. SLC7A8 also plays a role in basolateral (re)absorption of neutral amino acids.
Specifications

Specifications

SLC7A8
Monoclonal
0.65 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q9UHI5
Solute carrier family 7 (amino acid transporter light chain, L system) member 8, LAT2, LPI-PC1
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
23428
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
OTI4H10
Unconjugated
SLC7A8
Liquid
SLC7A8
Full length human recombit protein of SLC7A8 produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit