Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC8A3 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548456

Catalog No. PA548456

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Rapidly transports Ca(2+) during excitation-contraction coupling. Ca(2+) is extruded from the cell during relaxation so as to prevent overloading of intracellular stores.
Specifications

Specifications

SLC8A3
Polyclonal
PBS with 0.09% sodium azide
P57103
Sodium/calcium exchanger 3, Na(+)/Ca(2+)-exchange protein 3, Solute carrier family 8 member 3, SLC8A3, NCX3
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
6547
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
SLC8A3
Liquid
SLC8A3
KLH conjugated synthetic peptide between 585-614 amino acids from the C-terminal region of human SLC8A3.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit