Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC9A2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548427

Catalog No. PA548427

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Involved in pH regulation to eliminate acids generated by active metabolism or to counter adverse environmental conditions. Major proton extruding system driven by the inward sodium ion chemical gradient. Seems to play an important role in colonic sodium absorption.
Specifications

Specifications

SLC9A2
Polyclonal
PBS with 0.09% sodium azide
Q9UBY0
Sodium/hydrogen exchanger 2, Na(+)/H(+) exchanger 2, NHE-2, Solute carrier family 9 member 2, SLC9A2, NHE2
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
6549
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
SLC9A2
Liquid
SLC9A2
KLH conjugated synthetic peptide between 754-783 amino acids from the C-terminal region of human SLC9A2.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit