Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLCO6A1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP159898
Description
SLCO6A1 Polyclonal specifically detects SLCO6A1 in Human samples. It is validated for Western Blot.Specifications
SLCO6A1 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
Cancer/testis antigen 48, CT48Organic anion-transporting polypeptide I, Gonad-specific transporter, GSTOATP-I, MGC26949, OATP6A1Solute carrier family 21 member 19, OATPY, Organic anion-transporting polypeptide 6A1, SLC21A19, solute carrier organic anion transporter family member 6A1, solute carrier organic anion transporter family, member 6A1, testis-specific organic anion transporter | |
Rabbit | |
Protein A purified | |
RUO | |
133482 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 100μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot | |
1 mg/ml | |
Western Blot 1.0 ug/ml | |
Q86UG4 | |
SLCO6A1 | |
Synthetic peptides corresponding to SLCO6A1(solute carrier organic anion transporter family, member 6A1) The peptide sequence was selected from the N terminal of SLCO6A1. Peptide sequence CCNNIRCFMIFYCILLICQGVVFGLIDVSIGDFQKEYQLKTIEKLALEKS. | |
100 μL | |
Primary | |
Expected identity based on immunogen sequence: Human: 100%. | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction