Learn More
Aviva Systems Biology Corp SLFN12 Antibody - N-terminal region (ARP57095_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP57095_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLFN12. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SLFN12. Alternate names: FLJ10260, SLFN3ApplicationWB. Species reactivity: Cow, Human
Catalog No. 50-014-62077