Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLFNL1 Mouse anti-Human, Clone: OTI2E9, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA501967

Catalog No. 50-167-3062


Add to cart

Description

Description

SLFNL1 belongs to the Schlafen family. The function of the SLFNL1 protein remains unknown. There are four different isoforms.
Specifications

Specifications

SLFNL1
Monoclonal
1.01 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q499Z3
schlafen like 1
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
200172
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
OTI2E9
Unconjugated
SLFNL1
Liquid
SLFNL1
Full length human recombit protein of human SLFNL1 produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit