Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLFNL1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA565907
Description
Immunogen sequence: TRNMEFKRGSG EYLSLAFKHH VRRYVCAFLN SEGGSLLVGV EDSGLVQGIR CSHRDEDRAR LLVDSILQGF KPQIFPDAYT LTFIPVISTS ETS Highest antigen sequence identity to the following orthologs - mouse 95%, rat 94%.
SLFNL1 belongs to the Schlafen family. The function of the SLFNL1 protein remains unknown. There are four different isoforms.
Specifications
SLFNL1 | |
Polyclonal | |
Unconjugated | |
SLFNL1 | |
4933406A14Rik; schlafen like 1; schlafen-like 1; schlafen-like protein 1; SLFNL1; testicular tissue protein Li 173 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
200172 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q499Z3 | |
SLFNL1 | |
Recombinant Human SLFNL1. Recombinant protein control fragment (Product #RP-105005). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction