Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Laxco™ SLi6 PRO Series Inverted Compound Microscope
GREENER_CHOICESDP

The SLi6 PRO Series Inverted Compound Microscope is an upgradable trinocular microscope. Phase-contrast, fluorescent, and phase-contrast fluorescent options are available with complementing filters included.

Supplier:  Laxco™ SLI6PPH1

 View more versions of this product

Catalog No. SLI6P-PH1


Explore available promotions
Add to Cart
Request a Quote

Description

Description

  • Infinity corrected optical system
  • Trinocular 45° inclined head rotates 360°
  • Adjustable interpupillary distance of 50 mm to 75 mm with 100:100 image split
  • High eye-point, wide-field plan eyepiece (10x/22 mm FOV) with adjustable eyecups
  • Coarse and fine low-position coaxial focusing mechanism with tension adjustment and upper limit
  • N.A. 0.3 condenser with 72 mm working distance which can be removed to view specimens in large vessel
  • Quintuple nosepiece
  • Upgrade kits available (sold separately)
Specifications

Specifications

NA 0.3 (WD 72 mm)
Infinity LWD Plan Positive Phase Contrast Objective (4X, 10X, 20X)
5 years optical and mechanical, 1 year electrical from date of purchase
Fixed
LED
SLi6P-PH1
Inverted Phase-contrast Microscope
40X to 200X
Quintuple Nosepiece
Phase Slider, 45 mm Monochromatic Contrast Color Filter (Green), Light Balancing Daylight Color Temperature Transition Filter
WF 10X/22 mm
Trinocular
SLi6 PRO Series
High
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.