Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Slide-A-Lyzer™ Dialysis Cassettes, 10K MWCO, 3 mL Available on GSA/VA Contract for Federal Government customers only.

Remove buffer salts and contaminants and maximize sample recovery using this semi-permeable membrane with a 10000-dalton protein MW cut-off (MWCO).

$117.70 - $539.50


Products 3
Catalog Number Mfr. No. Description Capacity (Metric) Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Description Capacity (Metric) Quantity Price Quantity & Availability  
PI66380
Accessories View Documents
Thermo Scientific™
66380
Slide-A-Lyzer Dialysis Cassettes, 10K MWCO, 3mL 3mL 10 cassettes
Pack of 10 for $117.70
Add to cart
 
PI66381 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
66381
Slide-A-Lyzer Dialysis Cassettes, 10K MWCO, 3mL 3mL 5 x 10 cassettes
Pack of 5 for $539.50
Add to cart
 
PI66405
View Documents
Thermo Scientific™
66405
Slide-A-Lyzer Dialysis Cassette Trial Kit, 10K MWCO, 3mL 3mL 1 cassette kit This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description

Description

Thermo Scientific™ Slide-A-Lyzer Dialysis Cassettes (10K MWCO) facilitate simple and effective removal of buffer salts and small contaminants from proteins and other macromolecules that are larger than 10,000 daltons.

Available in four different cassette sizes to accommodate sample volumes between 0.1 and 30mL, these devices are easier to use than traditional dialysis tubing, providing both faster buffer exchange and more complete sample recovery. Slide-A-Lyzer Cassettes can be used for a wide range of applications including low molecular-weight contaminant removal, buffer exchange, desalting, equilibrium dialysis, and sample concentration.

The cassettes are constructed from two sheets of low-binding, regenerated-cellulose dialysis membrane that are hermetically sealed on either side of a silicone-like gasket inside an inert plastic frame. The membrane and cassette materials are compatible with most common laboratory chemicals and buffers. Liquid samples are easily added and removed by penetrating the self-sealing gasket with a hypodermic needle attached to a syringe. No knots, clips or caps are needed to seal the units and prevent leaking and sample-loss during the dialysis procedure.

Highlights:

  • Easy to use – no knots or clamps are needed; just inject sample into cassette and begin dialysis

  • Fast dialysis – flat cassette chamber with two membranes provides high surface-area to volume ratio that maximizes diffusion rate compared to cylindrical dialysis tubing

  • High recovery – rectangular cassette design maximizes recovery of entire sample volume via any one of the four corner injection ports

  • Four cassette sizes – select the cassette that best suits your sample volume

  • Color-coded – orange cassette frame specifies cassettes with the 10,000 MW pore size membrane

10K Dialysis Membrane Specifications:

  • Composition: Regenerated cellulose synthesized by the Viscose method; 21% glycerol, 0.05% sulfur, Trace heavy metals content
  • Thickness: 0.9 to 1.2mil (22.5 to 30µm)
  • Hydration Required Before Use: 30 seconds for small-volume samples

Item-Specific Features:

  • 30mL cassette has built-in air chamber for flotation

Includes:

Kits contain cassettes plus an equal number of syringes with 18-gauge needles and single-cassette float buoys

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit