Learn More
Thermo Scientific™ Slide-A-Lyzer™ G2 Dialysis Cassettes, 10K MWCO, 0.5 mL
Remove buffer salts and contaminants while maximizing sample recovery with these second-generation self-floating protein dialysis cassettes (10K MWCO).
Manufacturer: Thermo Scientific™ 87729
Description
Thermo Scientific™ Slide-A-Lyzer™ G2 10K MWCO Dialysis Cassettes provide maximum convenience, flexibility and performance for sample dialysis.
These second generation (G2) Slide-A-Lyzer Dialysis Cassettes are free-standing, self-floating and pipette-loadable units for dialysis. The single-use, disposable cassettes are available in five different sizes to accommodated samples between 0.1mL and 70mL. They offer maximum efficiency, convenience and sample protection in one package. Sample loading and removal are easily accomplished by using a serological pipette or hypodermic needle (optional) attached to a syringe. The built-in air chamber provides sample buoyancy and vertical orientation of the cassette during dialysis.The 10K molecular-weight cutoff (MWCO) cassettes use a cellulose membrane that is rated to completely retain proteins and other macromolecules that are larger than 10,000 daltons while allowing removal of buffer salts and small contaminants. The membrane is composed of low-binding regenerated cellulose, and the cassettes are manufactured using clean-room conditions.
Highlights:
Easy-loading – pipette-accessible for easy sample loading and retrieval
Self-floating – integrated air chambers eliminate the need for float buoys
Sturdy construction – ensures the highest possible sample integrity and protection
Superior design – thoroughly researched and tested to provide fast and consistent dialysis with maximum sample recovery
Multiple sizes – five cassette capacities to optimally match 0.1 to 70mL sample volumes
Versatile – ideal for removing low molecular-weight contaminants, performing buffer exchange and desalting
10K Dialysis Membrane Specifications:
- Composition: Regenerated cellulose synthesized by the Viscose method; 21% glycerol, 0.05% sulfur, Trace heavy metals content
- Thickness: 0.9 to 1.2mil (22.5 to 30μm)
- Hydration Required Before Use: 30 seconds for small-volume samples
Specifications
Store at room temperature | |
Cellulose Membrane, Dialysis | |
0.5 mL | |
Slide-A-Lyzer™ |
Buffer Exchange, Protein | |
Cassette | |
10 kDa | |
10 Cassettes |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.