Learn More
Thermo Scientific™ Slide-A-Lyzer™ G2 Dialysis Cassettes, 2K MWCO
Important Note: These G2 Dialysis Cassettes will be discontinued on June 1, 2023 and replaced by G3 Dialysis Cassettes, a new generation of cassettes. Consider switching to G3 Dialysis Cassettes. Remove salts and contaminants while maximizing sample recovery with these second-generation, self-floating protein dialysis cassettes.
Specifications
Content And Storage | Store at room temperature |
---|---|
Purification Target | Buffer Exchange, Protein |
Membrane | Cellulose Membrane, Dialysis |
Format | Cassette |
MWCO | 2.0 kDa |
Catalog Number | Mfr. No. | Sample Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Sample Volume (Metric) | Price | Quantity & Availability | |||||
PI87717
|
Thermo Scientific™
87717 |
0.5 mL |
Item Discontinued
This item has been discontinued by Fisher Scientific. Please Sign In to view product availability in your area. View alternate product |
N/A | |||||
PI87718
|
Thermo Scientific™
87718 |
3 mL |
Item Discontinued
This item has been discontinued by Fisher Scientific. Please Sign In to view product availability in your area. View alternate product |
N/A | |||||
PI87719
|
Thermo Scientific™
87719 |
15 mL |
Item Discontinued
This item has been discontinued by Fisher Scientific. Please Sign In to view product availability in your area. View alternate product |
N/A | |||||
PI87720
|
Thermo Scientific™
87720 |
30 mL |
Item Discontinued
This item has been discontinued by Fisher Scientific. Please Sign In to view product availability in your area. View alternate product |
N/A | |||||
PI87721
|
Thermo Scientific™
87721 |
70 mL |
Item Discontinued
This item has been discontinued by Fisher Scientific. Please Sign In to view product availability in your area. View alternate product |
N/A | |||||
Description
Slide-A-Lyzer G2 2K molecular-weight cut off (MWCO) dialysis cassettes (2K MWCO) provide maximum convenience, flexibility, and performance for sample dialysis. The cassettes use a cellulose membrane that retains proteins and other macromolecules that are larger than 2000 Daltons, while allowing removal of buffer salts and small contaminants. The cassette membrane is composed of low-binding regenerated cellulose. The membranes and cassettes are manufactured using clean-room conditions to ensure they are contaminant-free.
Slide-A-Lyzer G2 Dialysis Cassettes allow samples to be directly injected with a syringe or pipette through the unique sample port. The cassette design allows air to be removed from inside the cassette to maximize the surface area-to-volume ratio, providing better dialysis performance than dialysis tubing and tube-based systems. The sample is easily recovered through either the pipette or syringe sample ports.Features of Slide-A-Lyzer G2 Dialysis Cassettes (2K MWCO):
• Easy-loading—pipette-accessible for easy sample loading and retrieval
• Self-floating—integrated air chambers eliminate the need for float buoys
• Sturdy construction—ensures the highest possible sample integrity and protection
• Superior design—thoroughly researched and tested to provide fast and consistent dialysis with maximum sample recovery
• Multiple sizes—five cassette capacities to optimally match 0.5 to 70 mL sample volumes
• Versatile—ideal for removing low-molecular weight contaminants, performing buffer exchange, and desalting
Volume ranges:
• 0.5 mL = 0.25 to 0.75 mL
• 3 mL = 1 to 3 mL
• 15 mL = 5 to 15 mL
• 30 mL = 10 to 30 mL
• 70 mL = 25 to 70 mL
Specifications
Store at room temperature | |
Cellulose Membrane, Dialysis | |
2.0 kDa |
Buffer Exchange, Protein | |
Cassette | |
Slide-A-Lyzer™ |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.