Learn More
Thermo Scientific™ Slide and Block Cabinets
Form an integrated storage system with stackable Thermo Scientific™ Slide and Block Cabinets, constructed of steel and finished in solvent-proof paint.
Manufacturer:  Thermo Scientific™ 161003
 View more versions of this product
Catalog No. 16-100-3
  Check Availability
Add to cart
Specifications
Description
Form an integrated storage system with stackable Thermo Scientific™ Slide and Block Cabinets, constructed of steel and finished in solvent-proof paint.
Features
  • Steel construction
  • Solvent-proof paint
  • Sealed top and bottom to ensure dust-free storage
  • Universal plinth in black metal for Slide Safe and Block Bank
  • Optional Wheeled Plinth capable of supporting up to ten Slide Safes
Slide Safe
  • Contains fourteen drawers
  • Total capacity of 6000 microscope slides
  • Dimensions: 18.9 x 18.9 x 5.1" (480 x 480 x 130mm)
Block Bank
  • Contains eight drawers
  • Approximate total capacity of 3000 blocks
  • Dimensions: 18.9 x 18.9 x 10.8" (480 x 480 x 275mm)
Ordering Information:
Foam dividers for slide drawers (Cat. No. B1000152) available in packs of 12.
Description & Specifications
Specifications
3000 Blocks
18.9 in.
18.9 in.
10.8 in.
Block Bank; Green
Green
480mm
480mm
275mm
Block Bank