Thermo Scientific™ Slide Mailer and Folder
Hold up to 20 slides with the Thermo Scientific™ Slide Mailer and Folder.
Manufacturer: Thermo Scientific™ 58308
 View more versions of this product
Catalog No. 58-308
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Material Plastic
    Item Description Slide Folder
    Hold up to 20 slides with the Thermo Scientific™ Slide Mailer and Folder.
    Slide Mailer
    • Cardboard construction
    • Mailer holds two 3 x 1 in. (76 x 25mm) microscope slides
    • 3.125 x 4 in. (79 x 102mm) configuration conforms to U.S. Postal regulations
    • Cut-out windows allow viewing of patient identification
    • 24 per box, 30 boxes per case
    Slide Folder
    • Plastic construction
    • See-through flaps eliminate need to open the folder when searching for slides
    • Easy to clean
  • Description & Specifications
Related Products
Product Content Feedback