Learn More
Grainger SLIP-JOINT PLIERS 6-1/8"
Supplier:  Grainger 1UKJ4
Encompass_Preferred
Westward Slip Joint Pliers, 6-1/8 in. Overall Length, 1 in. Max. Jaw Opening, Handle Type: Ergonomic
Catalog No. NC1042849