Learn More
Brady™ Slip, Trip & Floor Obstacle Hazard Labels - Self-Sticking Polyester
Slip, Trip & Floor Obstacle Hazard Labels - Self-Sticking Polyester: Self-Sticking Polyester (B-302) is a flexible polymer plastic over-laminated with a clear plastic coating to protect graphics against abrasion, sunlight, chemicals.
$15.75
Products 2
Catalog Number Mfr. No. Display Text Price Quantity & Availability  
Catalog Number Mfr. No. Display Text Price Quantity & Availability  
19-112-894 View Documents Brady™
86305
WALK DON'T RUN Pack of 5 for $15.75
Add to cart
19-112-895 View Documents Brady™
86306
WATCH YOUR STEP Pack of 5 for $15.75
Add to cart
Description
Description
  • Weathers up to 8 years outdoors, even longer indoors, and withstands wide temperature variations (up to 230°F)
  • Flexible material conforms to most surfaces while protected graphics hold up to harsh environments, abrasion, and repeated spills and cleanings
  • Permanent, pressure sensitive adhesive forms a long-lasting bond to even the most difficult surfaces and shapes
  • Cold temperature adhesive allows the signage to be applied at 0°F (-18°C)