Learn More
Thermo Scientific™ SLIPFREE™ HPLC Column Single Connectors
Ensure leak-free, void-free column connections and optimize LC performance with universal, self-adjusting Thermo Scientific? SLIPFREE? HPLC Column Connectors.
Manufacturer:  Thermo Scientific™ 32520
 View more versions of this product
Catalog No. 03-150-801
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Description
Ensure excellent column connections and optimal HPLC performance with Thermo Scientific? SLIPFREE? HPLC Column Single Connectors. Compatible with all column end-fittings, these universal, self-adjusting connectors provide leak- and void-free column connections. Fingertight connections up to 10000psi ensure ease of use. They are ideal for supercritical fluid columns (SFC). SLIPFREE connectors are available in a variety of configurations and designs for a broad range of applications.