Learn More
Thermo Scientific™ SLIPFREE Ferrules Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ SLIPFREE Ferrules are for use with SLIPFREE connectors for HPLC columns.
Manufacturer:  Thermo Scientific™ 36023
 View more versions of this product
Catalog No. 03-170-504
  Check Availability
Add to cart
Description
Description
Thermo Scientific™ SLIPFREE Ferrules are for use with SLIPFREE connectors for HPLC columns.
  • Vespel ferrules replace the standard Vespel ferrules supplied with SLIPFREE column connectors
  • Kel-F and PEEK are offered for applications in which Vespel is not suitable
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
SLIPFREE™ connectors for HPLC columns
SLIPFREE connectors for HPLC columns
PEEK sealing ferrule
PEEK
PEEK