Learn More
Thermo Scientific™ SLIPFREE™ HPLC Column Double Connectors
Ensure leak-free, void-free column connections and optimize LC performance with self-adjusting Thermo Scientific? SLIPFREE? HPLC Column Double Connectors.
Manufacturer:  Thermo Scientific™ 32710
 View more versions of this product
Catalog No. 32-710
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Description
Ensure excellent column connections and optimal HPLC performance with Thermo Scientific? SLIPFREE? HPLC Column Double Connectors. Compatible with all column end-fittings, these universal, self-adjusting connectors provide leak- and void-free column connections. Fingertight connections up to 10000psi ensure ease of use. They are ideal for supercritical fluid columns (SFC). SLIPFREE connectors are available in a variety of configurations and designs for a broad range of applications.
Leak-Free, Void-Free Connections:
  • Pushes tubing and ferrule into end-fitting
  • Self adjusts to fit each new column connection
  • Fingertight connection up to 10000 psi
Flexible:
  • Fits all brands of columns
  • Available in single and double configurations
  • Available in a range of internal diameters to suit multiple applications
  • PEEK-collared and long-neck designs available for specific applications
Description & Specifications
Specifications
Double Long-Neck
0.02 in.
Double Long-Neck; 0.020in. ID x 10cm
Injector, Semi Preparative and Preparative Connection
10cm