Learn More
Thermo Scientific™ SLIPFREE™ Sample Loops Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ SLIPFREE Sample Loops feature a self-adjusting, leak-free design.
Manufacturer:  Thermo Scientific™ 32650
 View more versions of this product
Catalog No. 32-650
  Check Availability
Add to cart
Specifications
Description
Thermo Scientific™ SLIPFREE Sample Loops feature a self-adjusting, leak-free design.
  • Compatible with Rheodyne and Valco™ injectors
  • Long neck design
Description & Specifications
Specifications
Rheodyne and Valco Injectors
Sample Loop
0.020 in.
100?L
Liquid Chromatography
100uL volume; 0.020 in. ID; 50cm length
50cm