Learn More
Thermo Scientific™ SLIPFREE™ Sample Loops Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ SLIPFREE Sample Loops feature a self-adjusting, leak-free design.
Manufacturer:  Thermo Scientific™ 32699
 View more versions of this product
Catalog No. 32-699
  Check Availability
Add to cart
Specifications
Description
Thermo Scientific™ SLIPFREE Sample Loops feature a self-adjusting, leak-free design.
  • Compatible with Rheodyne and Valco™ injectors
  • Long neck design
Description & Specifications
Specifications
Rheodyne and Valco Injectors
Sample Loop
0.020 in.
250?L
Liquid Chromatography
250uL volume; 0.020 in. ID; 125cm length
125cm