Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FyterTech Nonwovens Slipfyter™ Traction Mat

Catalog No. p-7138805
Click to view available options
Capacity (English):
10.5 gal.
21 gal.

Slipfyter™ Traction Mats roll out easily for continuous coverage at entrances and walkways. No gaps, ripples or overlaps that can be slip, trip or fall hazards

  • Easy, durable solution to combat slips, trips and falls in a variety of locations
  • Withstand heavy foot and vehicle traffic, including fork trucks
  • Bottom layer sticks to the ground/surface with Cling technology
  • Peel up and re-position as needed
  • Durable top layer can be vacuumed, swept with broom or mopped
  • ADA compliant for wheelchairs and walkers
  • A fraction of the weight of rubber-backed mats: easing back strain when changing mats
  • Create custom-sized rugs for transition areas and problem spots where leaks or spills may be present

Specifications

Color Gray
Description Traction Rug
Capacity (English) 21 gal.
Capacity (Metric) 79 L
Material Needle-punch Polypropylene
Type Absorbent Rug
For Use With (Application) Floors, walkways, entryways
Packaging Quantity 1 Roll
Weight Capacity Medium-weight
Dimensions (L x W x H) 14 x 14 x 34.375 in.
Length (English) 14 in.
Length (Metric) 0.3556 m
Width (English) 14 in.
Width (Metric) 0.355 m
Height (English) 34.375 in.
Height (Metric) 0.87 m
Diameter (English) 14 in.
Diameter (Metric) 0.3556 m
Weight (English) 30 lb.
Weight (Metric) 16 kg
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.