Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Slit2 Antibody (710305), mFluor Violet 610 SE, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB5444MFV610
Description
Slit2 Monoclonal antibody specifically detects Slit2 in Human, Mouse samples. It is validated for Flow Cytometry, CyTOFSpecifications
| Slit2 | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Rat | |
| 0.1 mL | |
| Growth and Development, Neuronal Cell Markers, Neuroscience, Signal Transduction | |
| 9353 | |
| Store at 4C in the dark. |
| Flow Cytometry, CyTOF | |
| 710305 | |
| Flow Cytometry, CyTOF-ready | |
| FLJ14420, slit (Drosophila) homolog 2, slit homolog 2 (Drosophila), slit homolog 2 protein, Slit-2SLIL3 | |
| Chinese hamster ovary cell line CHO-derived recombinant mouse Slit2, Gln26-Gln900, Accession # Q9R1B9 | |
| RUO | |
| Primary | |
| Human, Mouse | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction