Learn More
SLIT2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA532586
Description
Heat-mediated antigen retrieval is recommended prior to staining, using a 1mM EDTA buffer, pH 8.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 10 min at room temperature. A suggested positive control is prostate adenocarcinoma or fetal kidney.
Slit homolog 2 proteins (SLIT2) are ligands for the Roundabout (Robo) receptors and act as molecule guidance cues in cell migration. Slit1 and Slit2 are essential for midline guidance in the forebrain by acting as repulsive signals preventing inappropriate midline crossing by axons projecting from the olfactory bulb. Slit2 protein is expressed in fetal lung and kidney, and adult spinal cord. Weak expression is also noticed in adult adrenal gland, thyroid, and trachea. SLIT2 is a target for repression by EZH2 and is up-regulated in prostate cancer.
Specifications
SLIT2 | |
Polyclonal | |
SLIT2 | |
SLIL3, Slit-2, slit homolog 2 protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9353 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
O94813 | |
SLIT2 | |
Synthetic peptide near C-terminus of human SLIT2 | |
500 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.