Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLIT2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | SLIT2 |
---|---|
Concentration | 0.19 mg/mL |
Applications | Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-2773
![]() ![]() |
Proteintech Group Inc
202171AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
The antibody is specific to SLIT2.
Slit homolog 2 proteins (SLIT2) are ligands for the Roundabout (Robo) receptors and act as molecule guidance cues in cell migration. Slit1 and Slit2 are essential for midline guidance in the forebrain by acting as repulsive signals preventing inappropriate midline crossing by axons projecting from the olfactory bulb. Slit2 protein is expressed in fetal lung and kidney, and adult spinal cord. Weak expression is also noticed in adult adrenal gland, thyroid, and trachea. SLIT2 is a target for repression by EZH2 and is up-regulated in prostate cancer.Specifications
SLIT2 | |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
O94813, Q9R1B9, Q9WVC1 | |
20563, 360272, 9353 | |
Peptide | |
Primary | |
-20°C | |
SLIT2 |
0.19 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
SLIL3, Slit 2, slit homolog 2 (Drosophila), Slit homolog 2 protein, SLIT2 | |
Slit2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title