Learn More
SLITRK1 Rabbit anti-Human, Mouse, DyLight™ 350, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus Biologicals NBP149541UV
Catalog No. NBP149541UV
Add to cart
Description
Description
SLITRK1 Polyclonal antibody specifically detects SLITRK1 in Human,Mouse samples. It is validated for Western Blot,Immunohistochemistry,Immunocytochemistry,Immunofluorescence.
Specifications
Specifications
SLITRK1
DyLight™ 350
50 mM sodium borate with 0.05% sodium azide
SLITRK1
A genomic peptide made to an internal region of the human Slitrk1 protein (within residues 250-400). [Swiss-Prot Q96PX8]
0.1 mL
Primary
114798
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
FLJ54428, KIAA1910KIAA0918, leucine rich repeat containing 12, Leucine-rich repeat-containing protein 12, LRRC12, RP11-395N17.1, SLIT and NTRK-like family, member 1, SLIT and NTRK-like protein 1, slit and trk like gene 1, TTM
Rabbit
IgG
Store at 4°C in the dark.
Polyclonal
Human, Mouse