Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ProSci Slitrk5 Peptide
SDP

Catalog No. 16919841
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-919-841 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-919-841 Supplier ProSci Supplier No. 4475P0050
Add to Cart
Add to Cart

Used for Blocking Activity

Slitrk5 peptide is used for blocking the activity of Slitrk5 antibody.

Specifications

Gene ID (Entrez) 26050
Species Human
Accession Number O94991
Common Name SLIT and NTRK-like family, member 5
Concentration 200 μg/mL
Content And Storage Store Slitrk5 peptide at -20°C, stable for one year.
Format Liquid
Formulation PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin and 0.02% sodium azide
For Use With (Application) Blocking Assay
Gene Symbol SLITRK5
Quantity 0.05 mg
Sequence 14 amino acids near the center of human Slitrk5.
Synonym LRRC11, bA364G4.2, KIAA0918, LRRC11, SLIT and NTRK-like protein 5, Leucine-rich repeat-containing protein 11
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.