Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLK Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549556

Catalog No. PA549556

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Mediates apoptosis and actin stress fiber dissolution.
Specifications

Specifications

SLK
Polyclonal
PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4
Q9H2G2, O54988, O08815
STE20-like serine/threonine-protein kinase, STE20-like kinase, hSLK, CTCL tumor antigen se20-9, STE20-related serine/threonine-protein kinase, STE20-related kinase, Serine/threonine-protein kinase 2, SLK, KIAA0204, STK2
Rabbit
IgG
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
20874, 54308, 9748
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
Unconjugated
SLK
Liquid
SLK
synthetic peptide corresponding to 1166-1191aa of human SLK
Affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit