Learn More
SLK Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA549878
Catalog No. PA549878
Add to cart
Description
Description
The antibody detects endogenous levels of total SLK protein.
Mediates apoptosis and actin stress fiber dissolution.
Specifications
Specifications
SLK
Polyclonal
Unconjugated
SLK
Liquid
SLK
Synthesized peptide derived from C-terminal of human SLK.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q9H2G2, O54988, O08815
KIAA0204, STK2
Rabbit
IgG
100 μL
-20°C
Primary
20874, 54308, 9748