Learn More
SLMAP Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA589784
Description
Immunogen sequence: IDTENLREEK DSEITSTRDE LLSARDEILL LHQAAAKVAS ERDTDIASLQ EELKKVRAEL ERWRKAASEY EKEITSLQNS FQLRCQQCED QQREEATRLQ GELEKLRKEW NALETECHSL KRENVLLSSE LQRQEKELHN SQKQSLELTS DLSILQMSRK ELENQVGSLK EQHLRDSADL KTLLSKAENQ AKDVQKEYEK TQTVLSELKL KFEMTEQEKQ SITDELKQCK NNLKLLREKG NNKPWPWMPM L; Positive Samples: Mouse heart, Rat heart; Cellular Location: Cell membrane, Cytoplasm, Single-pass type IV membrane protein, centrosome, cytoskeleton, microtubule organizing center, sarcolemma
Sarcolemmal membrane associated protein (SLMAP) is a tail-anchored membrane protein involved in the docking and membrane fusion process via protein-protein interactions. Deregulation of SLMAP expression plays an important role in type 2 diabetes, and changes in SLMAP expression are linked to hyperglycemia and diabetes. SLMAPs are components of cardiac membranes and have been identified as a novel regulator of cardiac function. There are several isoforms of SLMAPs with varying molecular size (35, 45, 63, and 83-91 kDa) as a result of alternative splicing. The short 35- kDa isoform is predomitly expressed in cardiac and slow-twitch skeletal muscle, while the 83- to 91- kDa isoform is ubiquitous.Specifications
SLMAP | |
Polyclonal | |
Unconjugated | |
Slmap | |
Slmap | |
Recombit fusion protein containing a sequence corresponding to amino acids 540-790 of human SLMAP | |
100 μL | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |
Western Blot | |
3.58 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P0C219, Q3URD3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
290533, 83997 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.